missing translation for 'onlineSavingsMsg'
Learn More

MUC-1 Ab-5 Hamster Monoclonal Antibody, Epredia™

Epredia™ MUC-1 Ab-5, Hamster Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia HM-1630-P1

 View more versions of this product

Product Code. 12638606

  • ONLINE EXCLUSIVE PRICE
    486.00 €
    408.73 € / 500µL
    Save 77.27 €
    15% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Muc1 is a heavily O-glycosylated transmembrane protein expressed on most secretory epithelium, including mammary gland and some hematopoietic cells. It is expressed abundantly in lactating mammary glands and overexpressed in >90% breast carcinomas and metastases. In normal mammary gland it is expressed in apical surface of glandular epithelium. In breast cancer Muc1 is overexpressed; is underglycosylated and the apical localization is lost. Muc1 is transcribed as a larger precursor which is cleaved to form a larger mucin like subunit (265-400kDa) and a smaller subunit (14-28kDa) noncovalently associated with each other. Transgenic Muc1 has been shown to associate with all four erbB receptors and localize with erbB1 (EGFR) in lactating glands1. Muc1 can act as a ligand for ICAM-1 on HUVEC cells; it can bind beta-catenin, GSK3beta and it associates with Grb2-SOS upon phosphorylation.

Host Species: Hamster

Clone: MH1 (CT2)

Isotype: IgG

Species Reactivity: Human and Mouse. Others not tested.

Epitope: aa 239-255

Immunogen: A synthetic peptide corresponding to aa 239-255 (SSLSYTNPAVAATSANL) from the cytoplasmic tail of MUC-1

Molecular Weight: 14-28kDa

Positive Control: MCF-7 cells; Breast CA

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

MUC-1 Ab-5
Monoclonal
200µg/mL
Hamster
500 μL
Cancer and Tumor Biology
Human, Mouse
IgG
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
MH1; same as CT2
Unconjugated
A synthetic peptide corresponding to aa 239-255 (SSLSYTNPAVAATSANL) form the cytoplasmic tail of MUC1
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.