missing translation for 'onlineSavingsMsg'
Learn More

Osteopontin Rabbit Polyclonal Antibody, Epredia™

Take advantage of a quality product that delivers accurate results with the Epredia™ Osteopontin, Rabbit Polyclonal Antibody.

Brand:  Epredia RB-9097-P

 View more versions of this product

Product Code. 12663457

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Osteopontin is a 34kDa extracellular matrix protein with a cell-binding domain. It was originally identified as a major component of the non-collagenous organic bone matrix however it has subsequently been demonstrated in a wide range of normal adult tissue and body fluids. It is a multifunctional protein involved in bone mineralization, cell adhesion, cell migration, chronic inflammatory disease and transformation. This antibody should prove useful in studying the expression of this matrix glycoprotein in tissue homeostasis. This antibody is suitable for staining of formalin-fixed, paraffin-embedded tissues.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Pig and Dog. Others not tested.

Epitope: Internal domain

Immunogen: A synthetic peptide derived from internal domain of human Osteopontin

Molecular Weight: 34kDa

Positive Control: Jurkat cells; Gastric CA or Osteosarcoma

Cellular Localization: Cytoplasmic This antibody is suitable for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Osteopontin
Polyclonal
200µg/mL
Rabbit
1 mL
Extracellular Matrix
Canine, Human
IgG
Western Blot
Polyclonal
Unconjugated
A synthetic peptide derived from internal domain of human Osteopontin
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.