missing translation for 'onlineSavingsMsg'
Learn More

Pan Cytokeratin Ab-1 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Epredia™ Cytokeratin, Pan Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia MS-343-PABX


 View more versions of this product

Product Code. 12681087

  • ONLINE EXCLUSIVE PRICE
    443.00 €
    398.25 € / 200µL
    Save 44.75 €
    10% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI <5.7) and basic (pI >6.0) subfamilies. The acidic keratins have molecular weights of 56.5, 55, 51, 50, 50', 48, 46, 45, and 40 kDa. The basic keratins have molecular weights of 65-67, 64, 59, 58, 56 and 52 kDa. Members of acidic and basic subfamilies are found together in pairs. The composition of keratin pairs varies with cell type, differentiation status and environment. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

Host Species: Mouse

Clone: AE1/AE3

Isotype: IgG1 + IgG1

Species Reactivity: Human, Monkey, Cow, Rabbit, Mouse, Rat and Chicken. Others not known.

Immunogen: Human epidermal keratin

Positive Control: Skin, Lung CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cytokeratin, Pan Ab-1
Monoclonal
1mg/mL
Mouse
200 μL
Cytoskeletal Protein
Bovine, Chicken, Human, Mouse, Non-human Primate, Rabbit, Rat
IgG1
Immunofluorescence, Immunohistochemistry (Paraffin)
AE1/AE3
Unconjugated
Human epidermal keratin
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MS-343-PABX: Pan Cytokeratin Ab-1 Mouse Monoclonal Antibody

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1

Hazard Statement
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P262-Do not get in eyes, on skin, or on clothing.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P320-Specific treatment is urgent (see on this label).
  • P321-Specific treatment (see on this label).
  • P391-Collect spillage.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

Supplemental information
  • MIXTURE LIST-Contains : 3(2H)-isothiazolone,5-chloro-2-methyl-with 2-methyl-3(2H)-isothiazolone

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.