missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Perforin Ab-2, Mouse Monoclonal Antibody

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states Epredia™ Perforin Ab-2, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1834-R7


 View more versions of this product

Product Code. 12610247

  • ONLINE EXCLUSIVE PRICE
    402.00 €
    337.66 € / 7mL
    Save 64.34 €
    16% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Perforin is a potent cytolytic pore-forming protein. It is a specific marker of functionally active cytotoxic T-lymphocytes (CTLs) and natural killer (NK) cells. Perforin is a key effector mechanisms in T cell-mediated cytotoxicity. It mediates cytolysis of target cells by membrane damage and apoptosis.

Host Species: Mouse

Clone: 5B10

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: C-terminal

Immunogen: Recombinant protein corresponding to C-terminal region of human perforin

Molecular Weight: 70kDa

Positive Control: Lymphoma

Cellular Localization: Granular and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Perforin Ab-2
Monoclonal
Unconjugated
Recombinant protein corresponding to C-terminal region of human perforin
IVD
Primary
Prediluted
Immunohistochemistry (Paraffin)
5B10
Mouse
7 mL
T-Cell
Human
IgG1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • PERFORIN AB-2 MOUSE MABREADY-TO-USE FOR IHC

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1A

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P362+P364-Take off contaminated clothing and wash it before reuse.

Supplemental information
  • MIXTURE LIST-Contains : hydrogen peroxide, 5-Chloro-2-methyl-3-isothiazolone, 2-Methyl-3-isothiazolone,sodium azide

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.