Learn More
Prohibitin (Mitochondrial Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ Prohibitin (Mitochondrial Marker) Ab-1, Mouse Monoclonal Antibody.
148.92 € - 882.30 €
Specifications
| Antigen | Prohibitin (Mitochondrial Marker) Ab-1 |
|---|---|
| Clone | II-14-10 |
| Classification | Monoclonal |
| Host Species | Mouse |
| Regulatory Status | RUO |
| Product Code | Brand | Quantity | Conjugate | Form | Concentration | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Conjugate | Form | Concentration | Price | Quantity & Availability | ||
|
12640727
|
Epredia
MS-261-B0 |
100 μL | Biotin | Concentrated | 200μg/mL | ||||
|
12650727
|
Epredia
MS-261-B1 |
500 μL | Biotin | Concentrated | 200μg/mL | ||||
|
12630727
|
Epredia
MS-261-B |
1 mL | Biotin | Concentrated | 200μg/mL | ||||
|
12670727
|
Epredia
MS-261-P0 |
100 μL | Unconjugated | Concentrated | 200μg/mL | ||||
|
12690727
|
Epredia
MS-261-P1ABX |
100 μL | Unconjugated | Concentrated | 1mg/mL | ||||
|
12600737
|
Epredia
MS-261-PABX |
200 μL | Unconjugated | Concentrated | 1mg/mL | ||||
|
12680727
|
Epredia
MS-261-P1 |
500 μL | Unconjugated | Concentrated | 200μg/mL |
514.00 € 429.84 € / 500µL Save 84.16 € 16% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
|
12660727
|
Epredia
MS-261-P |
1 mL | Unconjugated | Concentrated | 200μg/mL | ||||
|
12640737
|
Epredia
MS-261-R7 |
7 mL | Unconjugated | Prediluted | Prediluted | ||||
Description
Prohibitin is an evolutionarily conserved protein located in the inner membrane of mitochondria. Prohibitin appears to be a very reliable marker of mitochondria. Prohibitin shows antiproliferative activity and has been proposed to play a role in normal cell cycle regulation, replicative senescence, cellular immortalization, and tumor suppression.Host Species: Mouse
Clone: II-14-10
Isotype: IgG1
Species Reactivity: Human, Pig, Rat, Mouse, and Chicken. Others not known.
Epitope: Not determined
Immunogen: Purified recombinant rat prohibitin protein
Molecular Weight: 30kDa
Positive Control: MAD109 or PC12 cells, Heart
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
| Prohibitin (Mitochondrial Marker) Ab-1 | |
| Monoclonal | |
| RUO | |
| Chicken, Human, Mouse, Pig, Rat | |
| IgG1 |
| II-14-10 | |
| Mouse | |
| Apoptosis | |
| Purified recombinant rat prohibitin protein | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.