missing translation for 'onlineSavingsMsg'
Learn More

S100 Protein Ab-1 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ S100 Protein Ab-1, Mouse Monoclonal Antibody..

Brand:  Epredia MS-296-B

 View more versions of this product

Product Code. 12630857

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.

Host Species: Mouse

Clone: 4C4.9

Isotype: IgG2a

Species Reactivity: Human, Cow, Rat and Mouse. Others not known.

Epitope: Not determined

Immunogen: Purified bovine brain S100 protein

Molecular Weight: ∽22kDa (nonreduced); ∽11kDa (reduced)

Positive Control: Melanoma or Schwannoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

S100 Protein Ab-1
Monoclonal
200μg/mL
Mouse
1 mL
Neuroscience
Bovine, Human, Mouse, Rat
IgG2a
Immunohistochemistry (Paraffin)
4C4.9
Biotin
Purified bovine brain S100 protein
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.