Learn More
S100 Protein Ab-1 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ S100 Protein Ab-1, Mouse Monoclonal Antibody..
Brand: Epredia MS-296-B1
Description
S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.Host Species: Mouse
Clone: 4C4.9
Isotype: IgG2a
Species Reactivity: Human, Cow, Rat and Mouse. Others not known.
Epitope: Not determined
Immunogen: Purified bovine brain S100 protein
Molecular Weight: ∽22kDa (nonreduced); ∽11kDa (reduced)
Positive Control: Melanoma or Schwannoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| S100 Protein Ab-1 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 500 μL | |
| Neuroscience | |
| Bovine, Human, Mouse, Rat | |
| IgG2a |
| Immunohistochemistry (Paraffin) | |
| 4C4.9 | |
| Biotin | |
| Purified bovine brain S100 protein | |
| IVD | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.