Learn More
Epredia™ Lab Vision™ S100A4 Rabbit Polyclonal Antibody with BSA and azide
Provide accurate, reproducible results with the Epredia™ S100 A4, Rabbit Polyclonal Antibody.
Brand: Epredia™ RB-9411-P0
Description
S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.Host Species: Rabbit
Species Reactivity: Human. Others not tested.
Immunogen: Recombinant protein encoding full-length human S100 A4
Molecular Weight: 10-12kDa
Positive Control: Human melanoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
| S100 A4 | |
| Polyclonal | |
| Unconjugated | |
| Recombinant protein encoding full length human S100A4 | |
| RUO | |
| Primary | |
| Concentrated |
| Immunohistochemistry (Paraffin) | |
| 200µg/mL | |
| Rabbit | |
| 100 μL | |
| Neuroscience | |
| Human |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.