missing translation for 'onlineSavingsMsg'
Learn More

Thrombospondin (TSP) Ab-1 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Thrombospondin (TSP) Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia MS-418-P1

 View more versions of this product

Product Code. 12651377

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.

Host Species: Mouse

Clone: A4.1

Isotype: IgM

Species Reactivity: Human, Mouse, Rat, Cow and Chicken. Others not known.

Epitope: N-terminal half of the central stalk-like region of TSP, which is recovered as a 50kDa fragment after chymotryptic digestion of TSP

Immunogen: Calcium-replete, native purified human TSP from the supernatant of thrombin-activated platelets

Molecular Weight: ∽450kDa (nonreduced)

Positive Control: HUVEC cells

Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting (Nonreduced only)
  • Immunofluorescence
  • Blocks Binding of TSP-1 to TSP-Rec./CD36
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Thrombospondin (TSP) Ab-1
Monoclonal
200μg/mL
Mouse
500 μL
Angiogenesis
Bovine, Chicken, Human, Mouse, Rat
IgM
Flow Cytometry, Immunofluorescence, Western Blot
A4.1
Unconjugated
Calcium-replete, native purified human TSP from the supernatant of thrombin-activated platelets
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.