Learn More
Labconco™ Sample Blocks for RapidVap™ N2/48 Evaporation Systems
2514.66 € - 2551.87 €
Requires: Glassware
Products 2
Product Code Brand Holds Price Quantity & Availability  
Product Code Brand Holds Price Quantity & Availability  
11746419
View Documents
Labconco™
7482400
35 mL Tubes
ONLINE EXCLUSIVE PRICE
2850.00 €
2514.66 € / Pack of 1
Save 335.34 €
11% Off
View alternative products
Save Up to 
»
Estimated Shipment: 27-11-2026
Add to basket
 Restricted Item
11726649
View Documents
Labconco™
7482200
10 mL Tubes
ONLINE EXCLUSIVE PRICE
2765.00 €
2551.87 € / Pack of 1
Save 213.13 €
7% Off
View alternative products
Save Up to 
»
Estimated Shipment: 31-12-2026
Add to basket
 Restricted Item
Description
Description
  • Four different interchangeable aluminum accessory blocks, coated with PTFE
  • Sample blocks hold either 48 smaller (4.5 mL to 26 mL) or eight larger (450 mL) samples
  • Glassware for all blocks sold separately
†For tubes up to maximum capacity shown. Actual sample volume is 66–75% total tube capacity and depends on tube shape, height and vortex speed.