missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labetalol hydrochloride, MedChemExpress
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Labetalol hydrochloride is a mixed alpha/beta adrenergic antagonist that is used to treat high blood pressure.
Specifications
Specifications
| Chemical Name or Material | Labetalol hydrochloride |
| CAS | 32780-64-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C19H25ClN2O3 |
| Quantity | 100 mg |
| Synonym | AH-5158 hydrochloride Sch-15719W |
| Solubility Information | DMSO : ≥ 100 mg/mL (274.07 mM) ∣H2O : 20 mg/mL (54.81 mM; Need ultrasonic) |
| SMILES | O=C(N)C1=CC(C(O)CNC(C)CCC2=CC=CC=C2)=CC=C1O.[H]Cl |
| Molecular Weight (g/mol) | 364.87 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction