Learn More
SHP Steriltechnik AG Laboklav 25 Insert, 4 Levels
Product Code. 31908790
Change view
Click to view available options
For Use With (Equipment):
Laboklav 25
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
31908790 Laboklav 25 Each
1 options
Product Code. 31908790 Supplier SHP Steriltechnik AG Supplier No. 30.4220.004.025
Estimated Shipment: 07-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Stainless steel insert with four levels provides efficient use of chamber space for optimal loading and organisation during sterilisation.
  • Insert with 4 Levels for Laboklav 25
  • Stainless steel
  • WxHxD (mm): 190 x 426,5 x 180
Specifications
Capacity (Metric) Not Applicable
For Use With (Equipment) Laboklav 25
Model Insert, 4 Levels
Product Type Shelf
Sterility Non-sterile