Learn More
SHP Steriltechnik AG Laboklav 55 Insert, 3 Levels
Product Code. 31908763
Change view
Click to view available options
For Use With (Equipment):
Laboklav 55
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
31908763 Laboklav 55 Each
1 options
Product Code. 31908763 Supplier SHP Steriltechnik AG Supplier No. 30.0055.000.100
Estimated Shipment: 21-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Stainless steel insert with three levels, designed for the Laboklav 55 horizontal device, provides efficient use of chamber space for optimal loading and organisation during sterilisation.
  • Insert with 3 Levels for Laboklav 55 horizontal device
  • Stainless steel
  • Ø x D (mm): 360 x 400
Specifications
For Use With (Equipment) Laboklav 55
Model Insert, 3 Levels
Product Type Insert, 3 Levels