missing translation for 'onlineSavingsMsg'
Learn More

SHP Steriltechnik AG Laboklav 80 Wire Mesh Basket

Product Code. 31908750
Change view
Click to view available options
Capacity (Metric):
25 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
31908750 25 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31908750 Supplier SHP Steriltechnik AG Supplier No. 30.0290.145.600
Estimated Shipment: 13-01-2027
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Stainless steel basket designed for horizontal units ensures secure placement and optimal organisation of instruments during sterilisation.

  • Wire mesh basket for Laboklav 80
  • Stainless Steel
  • for horizontal units
  • WxHxD (mm): 290 x 145 x 600

Tekniske data

Capacity (Metric) 25 L
For Use With (Equipment) Autoclaves, Laboklav 80
Model Basket, Laboklav 80
Product Type Wire Mesh Basket
Sterility Non-sterile
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.