missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Labtainer™ 2D BioProcess Container (BPC), Aegis™ 5-14 film, three ports, Luer locks
GREENER_CHOICE

Product Code. 17890667
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1 L
2 L
2000 mL
5 L
10 L
20 L
50 L
Unit Size:
Pack of 10
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
17890667 2 L Pack of 10
17870607 50 mL Pack of 10
17880667 100 mL Pack of 10
17800677 250 mL Pack of 10
17890607 500 mL Pack of 10
17800617 1 L Pack of 10
17820677 2000 mL Pack of 10
17840607 5 L Pack of 10
17870667 10 L Pack of 10
17880607 20 L Pack of 10
17860667 50 L Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 17890667 Supplier Thermo Scientific™ Supplier No. SH31049.05
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Thermo Scientific Labtainer 2D BioProcess Containers (BPCs) are composed of Aegis 5-14 single-web, multi-layer film.

Features of the Labtainer 2D BPC include:
• Helps eliminate post-use cleaning steps required with re-useable containers and reduce cross-contamination risks
• Constructed in an ISO 7 cleanroom under cGMP conditions

Applications of the Labtainer 2D BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors
• Prepare and store buffer, culture media, and process liquid
• Collect chromatography feed and fractions
• Harvest, storage, and transport bulk drug product and bulk drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters—used for bioburden reduction; sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance ISO 7
Color Transparent
Disposable Single-use
For Use With (Application) Bioprocessing
Sterility Sterile
Tubing Style C-Flex
Film Description Aegis 5-14
Port Detail 3 ports
Quantity 2 L
Capacity (Metric) 2 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.