Learn More
Thermo Scientific™ Labtainer™ 2D BioProcess Container (BPC), three ports, MPCs, quick-connect
Description
Features of the Labtainer 2D BPC include:
• Helps eliminate post-use cleaning steps required with re-useable containers and reduce cross-contamination risks
• Constructed in an ISO 7 cleanroom under cGMP conditions
Applications of the Labtainer 2D BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors
• Prepare and store buffer, culture media, and process liquid
• Collect chromatography feed and fractions
• Harvest, storage, and transport bulk drug product and bulk drug precursors
Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters—used for bioburden reduction; sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Capacity (Metric) | 5 L |
| Certifications/Compliance | ISO 7 |
| Color | Transparent |
| Disposable | Single-use |
| For Use With (Application) | Bioprocessing |
| Sterility | Sterile |
| Tubing Style | C-Flex |
| Film Description | Aegis 5-14 |
| Port Detail | 3 ports |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.