missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, C-Flex 374 MPC insert and body connectors, injection site
Shop All Thermo Scientific ProductsDescription
This Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (including an injection site). A wide range of capacities, port configurations and tubing styles are available.
- Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
- All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions
Applications:
- Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
- Preparing and storing buffer, culture media, and process liquid
- Collecting chromatography feed and fraction
- Harvesting, storing, and transporting bulk drug product and drug precursors
Specifications
Specifications
| Capacity (Metric) | 10 L |
| Content And Storage | Store at room temperature. |
| Dimensions | 56.6 x 32.5 cm (22.3 x 12.8 in.) |
| Film Description | ASI™ 28 |
| Outer Packaging | One 4-mil. poly bag |
| Port Detail | 3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site) |
| Material | Ethyl Vinyl Acetate |
| Quantity | 25/Cs. |
| Shipping Condition | Room Temperature |
| Sterilization Method | Irradiated |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?