Learn More
Thermo Scientific™ Labtainer™ BioProcess Container (BPC), 100 mL with 2 Ports, Luer Lock, 2D BPC
Shop All Thermo Scientific Products

Description
The 100 mL Thermo Scientific™ Labtainer™ BPC, 2D system is composed of CX5-14 single-web, multi-layer film, and includes 2 edge ports with luer lock body and insert connections.
Features:
- Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
- All Labtainers are constructed in an ISO 7 cleanroom under cGMP conditions.
Applications of the Thermo Scientific Labtainer BPC include:
- Harvest from, feed into, and collect samples from bioreactors and fermentors.
- Buffer, culture media, and process liquid preparation and storage.
- Chromatography feed and fraction collection.
- Harvest, storage and transport of bulk drug product and bulk drug precursors.
Customize standard BPC systems for optimized single-use technology
Choose from the industry's largest component library – with over 20,000 unique components that include:
- Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
- Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
- Filters—used for bioburden reduction; sterilization grade filters and vent filters.
- Tubing—thermoplastic elastomers, platinum-cured silicone.
- Sample ports—septum-style and needle-free ports.
- Clamps—sliders, pinch, and crimp bands.
Specifications
Specifications
| Green Features | ISCC PLUS |
| Capacity (Metric) | 100 mL |
| Dimensions | 14.7 cm x 14.2 cm (5.8 in. x 5.6 in.) |
| Outer Packaging | Packages placed in strong cardboard cartons with product and lot identification |
| Protective Inner Packaging | Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area |
| Quantity | 10 per case |
| Sterilization Method | Gamma Irradiated |
| Tubing Line 1 | Line 1. Body luer lock connection, polypropylene. No Tubing |
| Tubing Line 2 | Line 2. Insert luer lock connection, polypropylene. No Tubing Line |
| Product Line | Labtainer |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.