Learn More
Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 2 ports, capsule filter and MPC insert
Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.
Brand: Thermo Scientific™ SS00163-I
Description
This Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 2 ports (1 membrane capsule filter and 1 MPC insert with pinch clamps). It is available in 50, 100 and 200L capacities.
- Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
- All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions
Applications:
- Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
- Preparing and storing buffer, culture media, and process liquid
- Collecting chromatography feed and fraction
- Harvesting, storing, and transporting bulk drug product and drug precursors
BPC port specifications:
- Line 1: 0.2μm PES membrane capsule filter + pinch clamp. Tubing: C-Flex 374, length: 91.4cm (36 in.), ID x OD: 9.5 x 15.9mm (0.38 x 0.63 in.)
- Line 2: MPC insert + cap, pinch clamp. Tubing: C-Flex 374, length: 91.
Specifications
| Store at room temperature. | |
| 2 Ports (1 Membrane Capsule Filter and 1 MPC Insert With Pinch Clamps) | |
| C-Flex, 91.4 cm (36 in.) | |
| Two 4-mil. poly bags | |
| Labtainer™ | |
| 5/Cs. | |
| Irradiated | |
| 42.5 in. | |
| 30 in. |
| 100 L | |
| Polyethylene | |
| ASI™ 26/77 | |
| Capsule filter and MPC insert + cap | |
| Protective material around rough areas | |
| Room Temperature | |
| 108 x 76.2 cm (42.5 x 30 in.) | |
| 108 cm | |
| 76.2 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.