missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 2 ports, capsule filter and MPC insert

Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.

POA

Specifications

Content And Storage Store at room temperature.
Includes 2 Ports (1 Membrane Capsule Filter and 1 MPC Insert With Pinch Clamps)
Material Polyethylene
Tubing Style C-Flex, 91.4 cm (36 in.)
Film Description ASI™ 26/77
View More Specs

Products 3
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15979926
View Documents
Thermo Scientific™
SS00162-I
50 L
POA
5 units
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

15989926
View Documents
Thermo Scientific™
SS00163-I
100 L
POA
5 units
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

15999926
View Documents
Thermo Scientific™
SS00164-I
200 L
POA
5 units
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

This Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 2 ports (1 membrane capsule filter and 1 MPC insert with pinch clamps). It is available in 50, 100 and 200L capacities.

  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications:

  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquid
  • Collecting chromatography feed and fraction
  • Harvesting, storing, and transporting bulk drug product and drug precursors

BPC port specifications:

  • Line 1: 0.2μm PES membrane capsule filter + pinch clamp. Tubing: C-Flex 374, length: 91.4cm (36 in.), ID x OD: 9.5 x 15.9mm (0.38 x 0.63 in.)
  • Line 2: MPC insert + cap, pinch clamp. Tubing: C-Flex 374, length: 91.

Specifications

Specifications

Store at room temperature.
Polyethylene
ASI™ 26/77
Capsule filter and MPC insert + cap
Protective material around rough areas
Room Temperature
2 Ports (1 Membrane Capsule Filter and 1 MPC Insert With Pinch Clamps)
C-Flex, 91.4 cm (36 in.)
Two 4-mil. poly bags
Labtainer™
5/Cs.
Irradiated
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.