missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, capsule filter, MPC insert, injection port

Product Code. 17810807 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
10 L
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17810807 10 L Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17810807 Supplier Thermo Scientific™ Supplier No. SS00160I
  • ONLINE EXCLUSIVE PRICE
    1975.00 €
    1637.83 € / Pack of 10
    Save 337.17 €
    17% Off
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.

This Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 3 ports (1 membrane capsule filter, MPC insert, pinch clamps, and an injection site). It is available in 1, 5, 10 and 20L capacities.

  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications:

  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquid
  • Collecting chromatography feed and fraction
  • Harvesting, storing, and transporting bulk drug product and drug precursors

BPC port specifications:

  • Line 1: 0.2μm PES membrane capsule filter + pinch clamp. Tubing: TPE 374, length: 30.5cm (12 in.), ID x OD: 9.5 x 15.9mm (0.38 x 0.63 in.)
  • Line 2: MPC insert + cap, pinch clamp. Tubing: TPE 374, length: 91.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Includes 3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site)
Material Polyethylene
Tubing Style TPE
Film Description ASI™ 26/77
Outer Packaging Two 4-mil. poly bags
Port Detail Capsule filter and MPC insert + cap
Product Line Labtainer
Protective Inner Packaging Protective material around rough areas
Quantity 15/Cs.
Shipping Condition Room Temperature
Sterilization Method Irradiated
Dimensions (L x W x H) 32 x 56.5 cm (12.6 x 22.3 in.)
Width (English) 22.3 in.
Product Type Bioprocess Container
Capacity (Metric) 10 L
Length (Metric) 32 cm
Width (Metric) 56.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.