Learn More
Lactase Antibody (CL14164) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26848DL594
Product Code. 30297896
  • 770.00 € / 0.10mL
Description
Lactase Monoclonal antibody specifically detects Lactase in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lactase
Monoclonal
DyLight 594
EC 3.2.1, LAC, lactase, Lactase-glycosylceramidase, lactase-phlorizin hydrolase, LPH1, LPHlactase-phlorizin hydrolase-1
This antibody was developed using a synthetic peptide derived from P09848, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cellular Signaling, metabolism
3938
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry
CL14164
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified