missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Lactobionic acid, 97%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Lactobionic acid is an aldonic acid generated from lactose oxidation that has antioxidant, chelating, and moisture-retaining properties
- Lactobionic acid can chelate metal ions and reduce oxidative damage
- Can be used in the synthesis of nanomaterials and drug-delivery nanoparticles
- Has been shown to enhance solubility of macrolide antibiotics
Applications
- Laboratory studies have show that lactobionic acid application reduced atopic dermatitis in a mouse model of skin disease
- Lactobionic acid has been found to inhibit galectin-1 in rats
- Has been used in nanotechnology to improve the solubility of chitosan hydrogels or as a nanoparticle conjugate to promote targeting of anti-cancer therapies to tumors
Specifications
Specifications
| Color | White |
| Loss on Drying | 5% max. |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97% |
| Packaging | Plastic bottle |
| Quantity | 25 g |
| Beilstein | 31, 415 |
| Merck Index | 15, 5391 |
| Specific Rotation Condition | +22.80° (20°C c=10,H2O) |
| Specific Rotation | After 4 hrs.: +22.8° to +28.8° (20°C, 589nm) (c=10, water) |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction