missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lactoferrin Antibody Pair [HRP]
Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Lactoferrin.
Specifications
Specifications
| Assay Range | 78.125 pg/mL |
| Assay Sensitivity | 78.125-5000 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Lactoferrin. |
| Target Species | Human |
| For Use With (Application) | Sandwich ELISA |
| Gene Alias | EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin |
| Gene ID (Entrez) | 4057 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction