missing translation for 'onlineSavingsMsg'
Learn More

Lactosyl Maprotiline(>90%), TRC

Product Code. 30477640
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477640 25 mg 25mg
30476169 10 mg 10mg
30465854 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30477640 Supplier Toronto Research Chemicals Supplier No. TRCL16528025MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lactosyl Maprotiline
Analyte or Component Names Lactosyl Maprotiline
Shipping Condition Room Temperature
Molecular Formula C32 H43 N O10
InChI Formula InChI=1S/C32H43NO10/c1-33(14-6-12-32-13-11-17(18-7-2-4-9-20(18)32)19-8-3-5-10-21(19)32)30-27(39)26(38)29(23(16-35)41-30)43-31-28(40)25(37)24(36)22(15-34)42-31/h2-5,7-10,17,22-31,34-40H,6,11-16H2,1H3/t17?,22?,23?,24-,25?,26?,27?,28?,29+,30+,31-,32?/m0/s1
Solution Type Neat
SMILES CN(CCCC12CCC(c3ccccc13)c4ccccc24)[C@@H]5OC(CO)[C@@H](O[C@@H]6OC(CO)[C@H](O)C(O)C6O)C(O)C5O
IUPAC Name (2S,5R)-2-[(3S,6R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[methyl-[3-(1-tetracyclo[6.6.2.02,7.09,14]hexadeca-2(7),3,5,9(14),10,12-hexaenyl)propyl]amino]oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 601.684
Quantity 25 mg
Formula Weight 601.289
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.