missing translation for 'onlineSavingsMsg'
Learn More

LAD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16895654 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16895654 20 μL 20µL
16885654 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16895654 Supplier Proteintech Supplier No. 161361AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunohistochemical staining of 16136-1-AP shows strong extracellular matrixpositivity in human testis. The predicted MW of this protein is 57 kDa. IP results of 16136-1-AP detected a 62 kDa band.

Anchoring filament protein which is a component of the basement membrane zone.

Specifications

Antigen LAD1
Applications Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LAD1
Gene Accession No. O00515, P57016
Gene Alias LAD, Lad 1, LAD1, LadA, ladinin 1, Linear IgA disease antigen
Gene Symbols LAD1
Host Species Rabbit
Immunogen LAD1 Fusion Protein Ag9159
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16763, 3898
Target Species Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.