missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ LAL Reagent Grade 96-Well Plates

Product Code. 11525769
Change view
Click to view available options
Quantity:
50/Pk.
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11525769 50/Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11525769 Supplier Lonza™ Supplier No. 25340

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Currently Unavailable: We have no information when this product will be available.
This item is not returnable. View return policy

Certified to contain <0.0005 EU/well endotoxin. Lonza™ LAL Reagent Grade 96-Well Plates are individually wrapped microplates designed for use with a variety of LAL assays in the detection of gram-negative bacterial endotoxins. They are compatible with a wide variety of plate readers.

Limulus Amebocyte Lysate (LAL) assays are used in the detection of gram-negative bacterial endotoxins. They are generally performed using an LAL reagent kit, a 96-well microplate and an incubating absorbance plate reader (and, often, appropriate detection software). These microplates are designed for use with:

  • QCL-1000™ Endpoint Chromogenic LAL Assay
  • Kinetic-QCL™ Kinetic Chromogenic LAL Assay
  • PYROGENT-5000™ Kinetic Turbidimetric LAL Assay
  • PryoGene™ rFC Assay
TRUSTED_SUSTAINABILITY

Specifications

Certified <0.0005 EU/well endotoxin
Format Standard
Description Individually wrapped
Array 8 x 12
Product Type LAL Reagent Grade Multi Well Plate
No. of Wells 96
For Use With (Application)
  • QCL-1000™ Endpoint Chromogenic LAL Assay
  • Kinetic-QCL™ Kinetic Chromogenic LAL Assay
  • PYROGENT-5000™ Kinetic Turbidimetric LAL Assay
  • PryoGene™ rFC Assay
Quantity 50/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.