missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lambda Light Chain Antibody (1-155-2), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-79124PEATT594
This item is not returnable.
View return policy
Description
Lambda Light Chain Monoclonal specifically detects Lambda Light Chain in Human samples. It is validated for Flow Cytometry.
Specifications
| Lambda Light Chain | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| IGLC1 | |
| Human IgA1 lambda myeloma protein | |
| 0.1 mL | |
| Primary | |
| The mouse monoclonal antibody 1-155-2 recognizes lambda light chains (22.5 kDa) of human immunoglobulin. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| 1-155-2 | |
| Flow Cytometry | |
| IGLC, immunoglobulin lambda constant group | |
| Mouse | |
| Protein A purified | |
| Adaptive Immunity, Immunology | |
| 3537.0 | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction