Learn More
Lamin A/C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10298-1-AP-150UL
Product Code. 16846593
  • 189.00 € / 150µL
Description
This antibody can recognize 4 isoforms of Lamin A/C.
The nuclear lamina consists of a two-dimensional matrix of proteins located next to the inner nuclear membrane. The lamin family of proteins make up the matrix and are highly conserved in evolution. During mitosis, the lamina matrix is reversibly disassembled as the lamin proteins are phosphorylated. Lamin proteins are thought to be involved in nuclear stability, chromatin structure and gene expression. Vertebrate lamins consist of two types, A and B. Alternative splicing results in multiple transcript variants. Mutations in this gene lead to several diseases: Emery-Dreifuss muscular dystrophy, familial partial lipodystrophy, limb girdle muscular dystrophy, dilated cardiomyopathy, Charcot-Marie-Tooth disease, and Hutchinson-Gilford progeria syndrome.
Specifications
Specifications
Lamin A/C
Polyclonal
Unconjugated
LMNA
70 kDa lamin, CDCD1, CDDC, CMD1A, CMT2B1, EMD2, FPL, FPLD, HGPS, IDC, lamin A, lamin A/C, LDP1, LFP, LGMD1B, LMN1, LMNA, LMNC, Prelamin A/C, PRO1, progerin
Rabbit
Antigen Affinity Chromatography
RUO
16905, 4000, 60374
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.45 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P02545, P48678, P48679
Lmna
Lamin A/C Fusion Protein Ag0408
150 μL
Primary
Human, Mouse, Rat