missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lamin B1 Antibody (CL3929) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59783B
This item is not returnable.
View return policy
Description
Lamin B1 Monoclonal antibody specifically detects Lamin B1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Lamin B1 | |
| Monoclonal | |
| Biotin | |
| ADLD, lamin B1, lamin-B1, LMN, LMN2, LMNB, MGC111419 | |
| This antibody was developed using a synthetic peptide derived from P20700, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Apoptosis, Cancer, Core ESC Like Genes, Loading Controls, Phospho Specific, Stem Cell Markers, Tumor Suppressors | |
| 4001 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL3929 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction