Learn More
Laminin alpha 2 Antibody (CL3450) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-46624UV
Product Code. 30288282
  • 737.00 € / 0.10mL
Description
Laminin alpha 2 Monoclonal antibody specifically detects Laminin alpha 2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Laminin alpha 2
Monoclonal
DyLight 350
Laminin M chain, laminin subunit alpha-2, laminin, alpha 2, Laminin-12 subunit alpha, Laminin-2 subunit alpha, LAMMLaminin-4 subunit alpha, Merosin heavy chain
This antibody was developed using a recombinant protein derived from P24043, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human, Mouse, Rat
Purified
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL3450
50mM Sodium Borate
Mouse
Protein A purified
RUO
3908
Store at 4⪚C in the dark.
IgG1