missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin alpha 2 Antibody (CL3450) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46624DL594
This item is not returnable.
View return policy
Description
Laminin alpha 2 Monoclonal antibody specifically detects Laminin alpha 2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Laminin alpha 2 | |
| Monoclonal | |
| DyLight 594 | |
| Laminin M chain, laminin subunit alpha-2, laminin, alpha 2, Laminin-12 subunit alpha, Laminin-2 subunit alpha, LAMMLaminin-4 subunit alpha, Merosin heavy chain | |
| This antibody was developed using a recombinant protein derived from P24043, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL3450 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 3908 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction