missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin alpha 5 Antibody (CL3118) [Alexa Fluor™ 647], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42391AF647
This item is not returnable.
View return policy
Description
Laminin alpha 5 Monoclonal antibody specifically detects Laminin alpha 5 in Human,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
Specifications
| Laminin alpha 5 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| KIAA0533, KIAA1907, laminin alpha-5 chain, laminin subunit alpha-5, laminin, alpha 5, Laminin-10 subunit alpha, Laminin-11 subunit alpha, Laminin-15 subunit alpha | |
| This antibody was developed using a recombinant protein derived from O15230, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human, Rat | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown | |
| CL3118 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 3911 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction