Learn More
Invitrogen™ Laminin alpha-5 Monoclonal Antibody (CL3118)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524649
Description
Immunogen sequence: LNLSSIILDV NQDRLTQRAI EASNAYSRIL QAVQAAEDAA GQALQQADHT WATVVRQGLV DRAQQLLANS TALEEAMLQE QQRLGLVWAA L Binds to an epitope located within the peptide sequence RQGLVDRAQQ as determined by overlapping synthetic peptides. Highest antigen sequence indentity to the following orthologs: Rat - 68%, Mouse - 62%.
Components of the extracellular matrix exert myriad effects on tissues throughout the body. In particular, the laminins, a family of heterotrimeric extracellular glycoproteins, affect tissue development and integrity in such diverse organs as the kidney, lung, skin, and nervous system. It is thought that laminins mediate the attachment, migration, and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components. Laminins function as heterotrimeric complexes of alpha, beta, and gamma chains, with each chain type representing a different subfamily of proteins. The protein encoded by this gene belongs to the alpha subfamily of laminin chains and is a major component of basement membranes. Two transcript variants encoding different isoforms have been found for this gene, but the full-length nature of one of them has not been determined.
Specifications
| Laminin alpha-5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15230 | |
| LAMA5 | |
| Recombinant protein fragment of Human LAMA5 (Product #RP-108178) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| CL3118 | |
| Unconjugated | |
| LAMA5 | |
| [a]5; AA408760; AA408762; AI853660; KIAA0533; KIAA1907; LAMA5; laminin alpha-5 chain; laminin subunit alpha 5; laminin subunit alpha-5; laminin, alpha 5; laminin-10 subunit alpha; Laminin-11 subunit alpha; laminin-15 subunit alpha; laminin-511; mKIAA0533; RP11-157P1.6 | |
| Mouse | |
| Protein A | |
| RUO | |
| 3911 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.