missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin Antibody (CL2968) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42384UV
This item is not returnable.
View return policy
Description
Laminin Monoclonal antibody specifically detects Laminin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Laminin | |
| Monoclonal | |
| DyLight 350 | |
| LAMA, Laminin A chain, laminin subunit alpha-1, laminin, alpha 1, Laminin-1 subunit alpha, Laminin-3 subunit alpha, PTBHS, S-LAM alpha, S-laminin subunit alpha | |
| This antibody was developed using a recombinant protein derived from P25391, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Neuroscience, Signal Transduction, Tumor Suppressors | |
| 284217 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL2968 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction