missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin gamma 1 Antibody (CL3196), Novus Biologicals™
Mouse Monoclonal Antibody
369.00 € - 529.00 €
Specifications
| Antigen | Laminin gamma 1 |
|---|---|
| Clone | CL3196 |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18643167
|
Novus Biologicals
NBP2-42395-25ul |
25 μL |
369.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18620397
|
Novus Biologicals
NBP2-42395 |
0.1 mL |
529.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Laminin gamma 1 Monoclonal antibody specifically detects Laminin gamma 1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Laminin gamma 1 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| P11047 | |
| 3915 | |
| IgG1 | |
| Protein A purified |
| CL3196 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Tumor Suppressors | |
| PBS (pH 7.2), 40% Glycerol | |
| LAMB2Laminin-7 subunit gamma, Laminin B2 chain, laminin subunit gamma-1, laminin, gamma 1 (formerly LAMB2), Laminin-1 subunit gamma, Laminin-10 subunit gamma, Laminin-11 subunit gamma, Laminin-2 subunit gamma, Laminin-3 subunit gamma, Laminin-4 subunit gamma, Laminin-6 subunit gamma, Laminin-8 subunit gamma, Laminin-9 subunit gamma, MGC87297, S-LAM gamma, S-laminin subunit gamma | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: ECREGFVGNRCDQCEENYFYNRSWPGCQECPACYRLVKDKVADHRVKLQELESLIANLGTGDEMVTDQAFEDRLKEAEREVMDLLREAQDVKDVDQNLMDRLQRVNNTLSSQISRLQNIRNTIEETGNLAEQARAHVENTERLIEI | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title