missing translation for 'onlineSavingsMsg'
Learn More

Lamivudine, MedChemExpress

Product Code. 16408196
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16408196 100 mg 100mg
16402076 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16408196 Supplier Medchem Express Supplier No. HYB0250
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lamivudine (BCH-189) is an orally active nucleoside reverse transcriptase inhibitor (NRTI). Lamivudine can inhibit HIV reverse transcriptase 1/2 and also the reverse transcriptase of hepatitis B virus. Lamivudine salicylate can penetrate the CNS.

Lamivudine (BCH-189) is an orally active nucleoside reverse transcriptase inhibitor (NRTI). Lamivudine can inhibit HIV reverse transcriptase 1/2 and also the reverse transcriptase of hepatitis B virus. Lamivudine salicylate can penetrate the CNS.

Specifications

Chemical Name or Material Lamivudine
Physical Form Powder
CAS 134678-17-4
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C8H11N3O3S
For Use With (Application) COVID-19-anti-virus
Synonym BCH-189
Solubility Information DMSO : 50 mg/mL (218.09 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (218.09 mM)
SMILES O=C1N=C(N)C=CN1[C@@H]2CS[C@H](CO)O2
Molecular Weight (g/mol) 229.26
Formula Weight 229.26
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.