missing translation for 'onlineSavingsMsg'
Learn More

Lanifibranor, MedChemExpress

Product Code. 16490297
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490297 50 mg 50mg
16464966 100 mg 100mg
16423948 10 mg 10mg
16406667 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16490297 Supplier Medchem Express Supplier No. HY104049
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lanifibranor is a pan peroxisome proliferator-activated receptor (PPAR) agonist with EC50s of 1.5, 0.87 and 0.21 μM for human PPARα, PPARσ and PPARγ, respectively.

Lanifibranor is a pan peroxisome proliferator-activated receptor (PPAR) agonist with EC50s of 1.5, 0.87 and 0.21 μM for human PPARα, PPARσ and PPARγ, respectively.

Spécification

Chemical Name or Material Lanifibranor
CAS 927961-18-0
Color Light Nude
Purity Grade Notes Research
Molecular Formula C19H15ClN2O4S2
Quantity 50 mg
Synonym IVA337
Solubility Information DMSO : 100 mg/mL (229.93 mM; Need ultrasonic and warming)
SMILES O=C(O)CCCC(N1S(=O)(C2=CC=C3N=CSC3=C2)=O)=CC4=C1C=CC(Cl)=C4
Molecular Weight (g/mol) 434.92
Formula Weight 434.92
Percent Purity 98.41%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Afficher plus Afficher moins

Research purposes only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.