missing translation for 'onlineSavingsMsg'
Learn More

Lanolin Anhydrous, Extra Pure, SLR, Fisher Chemical

Product Code. 10685302 Shop All Fisher Chemical Products
Change view
Click to view available options
500g, Plastic powder jar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10685302 500g, Plastic powder jar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10685302 Supplier Fisher Chemical Supplier No. L/0350/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 8006-54-0
Melting Point 38°C
Color Yellow
Packaging Plastic powder jar
MDL Number 81740
Quantity 500 g
Synonym dmt-2'o-tbdms-rc ac phosphoramidite, dmt-2'o-tbdms-rc ac phosphoramidite, configured for abi, dmt-2'o-tbdms-rc ac phosphoramidite, configured for mermade, ac-c-ce phosphoramidite, dmt-2'o-tbdms-rc ac amidite, dmt-2'o-tbdms-rc ac pharmadite r, dmt-2'o-tbdms-rc ac phosphoramidite, configured for ??kta r and oligopilot r, dmt-2'o-tbdms-rc ac phosphoramidite, configured for perkinelmer, configured for polygen, dmt-2'o-tbdms-rc ac phosphoramidite, configured for polygen, configured for perkinelmer
Physical Form Paste
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.