missing translation for 'onlineSavingsMsg'
Learn More

Lanolin, anhydrous, USP

Product Code. 10391391 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10391391 500 g Glass bottle 500g
10732784 2.5 kg Plastic drum 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10391391 Supplier Thermo Scientific Acros Supplier No. 413005000
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 8006-54-0

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8006-54-0
Synonym dmt-2'o-tbdms-rc ac phosphoramidite, dmt-2'o-tbdms-rc ac phosphoramidite, configured for abi, dmt-2'o-tbdms-rc ac phosphoramidite, configured for mermade, ac-c-ce phosphoramidite, dmt-2'o-tbdms-rc ac amidite, dmt-2'o-tbdms-rc ac pharmadite r, dmt-2'o-tbdms-rc ac phosphoramidite, configured for ??kta r and oligopilot r, dmt-2'o-tbdms-rc ac phosphoramidite, configured for perkinelmer, configured for polygen, dmt-2'o-tbdms-rc ac phosphoramidite, configured for polygen, configured for perkinelmer

Specifications

Alkalinity Passes Test
Acidity 2.0 ml 0.10 N NaOH max.
Melting Point 38°C to 44°C
Color White to Yellow
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 500 g
Merck Index 15, 5409
Physical Form Adhering Crystals or Powder
Chemical Name or Material Lanolin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.