missing translation for 'onlineSavingsMsg'
Learn More

Lanolin, Thermo Scientific Chemicals

Product Code. 11458726
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458726 250 g 250g
11448726 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11458726 Supplier Thermo Scientific Alfa Aesar Supplier No. A16902.30
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Lanolin, CAS # 8006-54-0, is a naturally occurring wax-like material that is obtained from wool of sheep.

Lanolin is used in a wide variety of creams, ointments and emollients. It is often used as protective baby skin treatment and lip balm products such as carmex. It acts as a lubricant, shoe polish, rust-preventative coatings and a starting material for the production of cholecalciferol using irradiation. It is used to soften and break baseball gloves. Further, It is used in various leather treatments by mixing with other ingredients like neatsfoot oil, beeswax and glycerine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

Lanolin is a waxy material that is composed of long chain esters and lanolin alcohols, acids, and polycarbons.

Application

• As a precursor of histidine, it can be used to enhance the biosynthesis of lovastatin by cultured Aspergillus terreus
• It can be used in stabilizing glass slides during worm-immobilization method

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8006-54-0
MDL Number MFCD00081740
Synonym dmt-2'o-tbdms-rc ac phosphoramidite, dmt-2'o-tbdms-rc ac phosphoramidite, configured for abi, dmt-2'o-tbdms-rc ac phosphoramidite, configured for mermade, ac-c-ce phosphoramidite, dmt-2'o-tbdms-rc ac amidite, dmt-2'o-tbdms-rc ac pharmadite r, dmt-2'o-tbdms-rc ac phosphoramidite, configured for ??kta r and oligopilot r, dmt-2'o-tbdms-rc ac phosphoramidite, configured for perkinelmer, configured for polygen, dmt-2'o-tbdms-rc ac phosphoramidite, configured for polygen, configured for perkinelmer

Specifications

Melting Point 38°C to 44°C
Quantity 250 g
Merck Index 14,5358
Solubility Information Soluble in ether,petroleum ether,chloroform and petroleum benzene; Sparingly soluble in ethanol; Insoluble in water.
Chemical Name or Material Lanolin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.