missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum Chloride, 7-Hydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Shop All J.T.Baker ProductsBeskrivelse
- For Flame Enhancement in Atomic Absorption Spectroscopy
Tekniske data
Tekniske data
| Packaging | Wide Mouth Amber Glass Bottle |
| Quantity | 100 g |
| Formula Weight | 371.38 |
| Grade | BAKER ANALYZED™ ACS Reagent |
| Chemical Name or Material | Lanthanum Chloride, 7-Hydrate |
Sikkerhed og håndtering
Produktidentifikator
- Lanthanum (III) chloride heptahydrate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contain: Lanthanum (III) chloride heptahydrate
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?