missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum Nitrate Releasing Agent: 1% La (from oxide) in 5% HNO3, VHG™

Product Code. 30003170
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003170 500 mL 500mL
30003064 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 30003170 Supplier VHG Labs Supplier No. VHGMLAN1P500
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Lanthanum Nitrate Releasing Agent: 1% La (from oxide) in 5% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lanthanum
CAS 7439-91-0
Methods AA/GFAA
Analyte or Component Names Lanthanum
Shipping Condition Room Temperature
Molecular Formula La
Synonym La
InChI Formula InChI=1S/La
SMILES [La]
IUPAC Name lanthanum
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 138.906
Quantity 500 mL
Formula Weight 138.906 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.