missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LanthaScreen™ Elite Tb-anti-HIS Antibody

LanthaScreen Elite Terbium-labeled anti-His-tag antibodies are used to indirectly label proteins tagged with 6XHis.

Brand:  Invitrogen™ PV5895

 View more versions of this product

Product Code. 10326318

  • 17000.00 € / 1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

LanthaScreen Elite Terbium-labeled anti-His-tag antibodies are used to indirectly label proteins tagged with 6XHis. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Tb chelates negatively impacts protein activity. The Elite Tb-anti-HIS-tag antibody is a specialized antibody with a broader HIS-tag binding performance than our traditional Tb-anti-HIS-tag antibody.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Kinase Assay
Tb (Terbium)/Lanthanide Chelates
150kDa
RUO
Primary
Elite Terbium-labeled anti-HIS antibody in HEPES buffered saline. Store at -20°C.
340/490, 520 nm
LanthaScreen™
0.53mg/mL
LanthaScreen™ Elite Tb-anti-HIS Antibody, 1mg, 0.53mg/mL Concentration, FITC (Fluorescein), Tb (Terbium) Conjugate, 340/490nm Excitation, 520nm Emission, 10mM HEPES (pH 7.5), 137mM NaCl and 2.7mM KCl (HEPES Buffered Saline) Storage Buffer, Used for
1 mg
Target and Lead Identification and Validation
1 Each
Frozen
Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.