missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ Tb-anti-AKT [pSer473] Antibody

Product Code. 10257074
Change view
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10257074 1 mg Each
10145404 25 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10257074 Supplier Thermo Scientific™ Supplier No. PV5124

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

AKT is phosphorylated at Ser473 by the rapamycin-insensitive mTor complex (mTORC2).

  • AKT is phosphorylated at Ser473 by the rapamycin-insensitive mTor complex (mTORC2)
  • For use in combination with the LanthaScreen™ AKT HEK293E cell line (K1516) to evaluate inhibitors in medium to high throughput screening applications
TRUSTED_SUSTAINABILITY

Specifications

Conjugate Tb (Terbium)/Lanthanide Chelates
Quantity 1 mg
Content And Storage Terbium-labeled pAKT (pSer473) antibody in HEPES buffered saline. Store at -20°C.
Excitation/Emission 340/490, 520
Reactivity Human
Shipping Condition Dry Ice
Readout End Point
Reactivity abrv Hu
Target Gene Ser/Thr Kinases
Product Line LanthaScreen

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.