Learn More
Invitrogen™ LanthaScreen™ Tb-anti-phospho-p53 [pSer15] Antibody, 25 μg
The p53 tumor suppressor protein is an essential component of many different signaling pathways.
Brand: Invitrogen™ PV5128
Description
The p53 tumor suppressor protein is an essential component of many different signaling pathways. The LanthaScreen™ antibody detects the phosphorylation of p53 at Ser15 by a range of kinases including DNA-PK, ATM, and ATR.
For use in combination with the Fluorescein-p53 (Ser15) peptide substrate (Cat. No. PV5132) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
| Ser/Thr kinases | |
| Tb (Terbium)/Lanthanide Chelates | |
| 25 μg | |
| Target and Lead Identification and Validation | |
| 1 Each | |
| Phosphorylation of p53 | |
| Human | |
| End Point | |
| Ser/Thr Kinases |
| Monoclonal | |
| 150kDa | |
| RUO | |
| Primary | |
| Terbium-labeled p53 (pSer15) antibody in HEPES buffered saline. Store at -20°C. |
|
| 340/490, 520 nm | |
| Dry Ice | |
| Hu | |
| LanthaScreen |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.