missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAP1 Antibody (RL13), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-2737C
This item is not returnable.
View return policy
Description
LAP1 Monoclonal antibody specifically detects LAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| LAP1 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| RL13 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| lamina-associated polypeptide 1B, torsin A interacting protein 1 | |
| Pore complex-lamina fraction isolated from rat liver nuclear envelopes. | |
| 0.2 mL | |
| Apoptosis, Immunology, Innate Immunity, Zinc Finger | |
| 208263 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction