missing translation for 'onlineSavingsMsg'
Learn More

β-Lapachone, MedChemExpress

Product Code. 16485748
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16485748 10 mg 10mg
16481907 50 mg 50mg
16456226 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16485748 Supplier Medchem Express Supplier No. HY13555
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress β-Lapachone (ARQ-501;NSC-26326) is a naturally occurring O-naphthoquinone, acts as a topoisomerase I inhibitor, and induces apoptosis by inhibiting cell cycle progression.

β-Lapachone (ARQ-501

Specifications

Chemical Name or Material β-Lapachone
CAS 4707-32-8
Color Brown
Purity Grade Notes Research
Molecular Formula C15H14O3
Quantity 10 mg
Synonym ARQ-501 NSC-26326
Solubility Information DMSO : 25 mg/mL (103.19 mM; Need ultrasonic) ∣Ethanol : 8.33 mg/mL (34.38 mM; Need ultrasonic)
SMILES O=C(C1=C2OC(C)(C)CC1)C(C3=C2C=CC=C3)=O
Molecular Weight (g/mol) 242.27
Formula Weight 242.27
Percent Purity 99.1%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.