missing translation for 'onlineSavingsMsg'
Learn More

β-Lapachone, MedChemExpress

Product Code. 16481907
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16481907

Brand: Medchem Express HY13555

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress β-Lapachone (ARQ-501;NSC-26326) is a naturally occurring O-naphthoquinone, acts as a topoisomerase I inhibitor, and induces apoptosis by inhibiting cell cycle progression.

β-Lapachone (ARQ-501

Specifications

Chemical Name or Material β-Lapachone
CAS 4707-32-8
Color Brown
Purity Grade Notes Research
Molecular Formula C15H14O3
Quantity 50 mg
Synonym ARQ-501 NSC-26326
Solubility Information DMSO : 25 mg/mL (103.19 mM; Need ultrasonic) ∣Ethanol : 8.33 mg/mL (34.38 mM; Need ultrasonic)
SMILES O=C(C1=C2OC(C)(C)CC1)C(C3=C2C=CC=C3)=O
Molecular Weight (g/mol) 242.27
Formula Weight 242.27
Percent Purity 99.1%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.