missing translation for 'onlineSavingsMsg'
Learn More

Lapatinib ditosylate, MedChemExpress

Product Code. 16461949
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16461949 1 g 1g
16431936 500 mg 500mg
16454047 50 mg 50mg
16427896 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16461949 Supplier Medchem Express Supplier No. HY50898A
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Lapatinib ditosylate (GW572016 ditosylate) is a potent inhibitor of the ErbB-2 and EGFR tyrosine kinase domains with IC50 values against purified EGFR and ErbB-2 of 10.2 and 9.8 nM, respectively.

Lapatinib ditosylate (GW572016 ditosylate) is a potent inhibitor of the ErbB-2 and EGFR tyrosine kinase domains with IC50 values against purified EGFR and ErbB-2 of 10.2 and 9.8 nM, respectively.

Specifications

Chemical Name or Material Lapatinib ditosylate
CAS 388082-77-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C43H42ClFN4O10S3
Quantity 1 g
Synonym GW572016 ditosylateGW2016 ditosylate
Solubility Information DMSO : 125 mg/mL (135.07 mM; Need ultrasonic)
SMILES O=S(CCNCC1=CC=C(C2=CC=C3C(C(NC4=CC(Cl)=C(C=C4)OCC5=CC(F)=CC=C5)=NC=N3)=C2)O1)(C)=O.O=S(C6=CC=C(C)C=C6)(O)=O.O=S(C7=CC=C(C)C=C7)(O)=O
Molecular Weight (g/mol) 925.46
Formula Weight 925.46
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.